Glassy carbon rod, 7mm (0.28in) dia, type 1, Thermo Scientific Chemicals
Glassy carbon rod, 7mm (0.28in) dia, type 1, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glassy carbon rod, 7mm (0.28in) dia, type 1, Thermo Scientific Chemicals

Used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used as electrical conductor ion-selective membrane.
Have Questions?
Change viewbuttonViewtableView
Quantity:
200 mm
50 mm
100 mm
Catalog number 038012.DR
also known as 038012-DR
Price (USD)/ Each
686.65
Online exclusive
763.00 
Save 76.35 (10%)
-
Add to cart
Quantity:
200 mm
Request bulk or custom format
Price (USD)/ Each
686.65
Online exclusive
763.00 
Save 76.35 (10%)
Add to cart
Glassy carbon rod, 7mm (0.28in) dia, type 1, Thermo Scientific Chemicals
Catalog number038012.DR
Price (USD)/ Each
686.65
Online exclusive
763.00 
Save 76.35 (10%)
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialGlassy carbon rod
Recommended StorageAmbient temperatures
EINECS Number231-153-3
TSCAYes
PackagingTube
View more
Glassy carbon rod is used as an electrode material in electrochemistry. It is also used as high temperature crucibles. Used as a component of some prosthetic devices. Used as electrical conductor ion-selective membrane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only