Thermo Scientific Chemicals

Glassy carbon splinter powder, 80 to 200μm, type 2, Thermo Scientific Chemicals

Catalog number: 038013.30
250 g, Each
Thermo Scientific Chemicals

Glassy carbon splinter powder, 80 to 200μm, type 2, Thermo Scientific Chemicals

Catalog number: 038013.30
250 g, Each
Quantity
Catalog number: 038013.30
also known as 038013-30
Price (USD)
Quantity
-
Have Questions?
Assay from Supplier's CofA
>99.9%
Appearance (Color)
Black
Form
Splinter powder, 80-200 micron (Glassy Carbon type-2, maximimum service temperature 3000?C).

Description

Glassy carbon powder is widely used as an electrode material in electrochemistry. It is used to make carbon paste electrodes in electrochemistry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text