Thermo Scientific Chemicals

Glassy carbon plate, 1mm (0.04in) thick, type 1, Thermo Scientific Chemicals

Catalog number: 038024.FI
50 x 50 mm, Each
Thermo Scientific Chemicals

Glassy carbon plate, 1mm (0.04in) thick, type 1, Thermo Scientific Chemicals

Catalog number: 038024.FI
50 x 50 mm, Each
Quantity
Catalog number: 038024.FI
also known as 038024-FI
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-44-0
IUPAC Name
carbon
Molecular Formula
C
InChI Key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES
[C]
Appearance (Color)
Black
Assay from Supplier's CofA
>99.9%
Form
Plate, 1mm (0.04in) thick (Glassy Carbon type-1, maximimum service temperature 1000?C).

Description

Used in casting, smelting of metal, hard alloys. Glassy carbon is widely used as an electrode material in electrochemistry, for high temperature crucibles and as a part of some prosthetic devices.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text