Thermo Scientific Chemicals

Glassy carbon plate, 4mm (0.16in) thick, type 1, Thermo Scientific Chemicals

Catalog number: 038026.GH
100 x 100 mm, Each
Thermo Scientific Chemicals

Glassy carbon plate, 4mm (0.16in) thick, type 1, Thermo Scientific Chemicals

Catalog number: 038026.GH
100 x 100 mm, Each
Quantity
Catalog number: 038026.GH
also known as 038026-GH
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7440-44-0
IUPAC Name
carbon
Molecular Formula
C
InChI Key
OKTJSMMVPCPJKN-UHFFFAOYSA-N
SMILES
[C]
Appearance (Color)
Black
Total Metal Impurities
≤0.1% (ash content)
Form
Plate, 4mm (0.16in) thick (Glassy Carbon type-1, maximimum service temperature 1000°C).

Description

Glassy carbon plate, is used as fuel cell and Due to their specific surface orientation, glassy carbon is employed as an electrode material for the fabrication of sensors. It is also used in mechanical applications such as when used as a structural component of a furnace. It is also used as also used for certain thin film deposition experiments. Chemically modified glasssy electrodes have been employed for the analysis of organic molecules (viz., Paracetamol, aspirin, caffeine, phenol, catechol, resorcinol, hydroquinone, dopamine, L-dopa, epinephrine, nor epinephrine, methyl parathion, ethyl parathion, venlafaxine, desvenlafaxine, imipramine, trimipramine, desipramine etc.) as well as metal ions (bismuth, antimony etc.)

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text