Palladium, 3% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Palladium, 3% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Palladium, 3% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
100 g
Catalog number 038296.14
also known as 038296-14
Price (USD)/ Each
-
Quantity:
25 g
Request bulk or custom format
Specifications
Chemical Name or MaterialPalladium
Name Note3% on activated carbon powder, Standard, reduced, nominally 50% water wet
CAS7440-05-3
Health Hazard 1H228
Health Hazard 3P210-P240-P241-P280-P370+P378q
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only