Thermo Scientific Chemicals
Palladium, 5% on activated carbon powder, Type A401102-5, eggshell unreduced, nominally 50% water wet, Thermo Scientific Chemicals
Catalog number: 038299.06
5 g, Each
Thermo Scientific Chemicals
Palladium, 5% on activated carbon powder, Type A401102-5, eggshell unreduced, nominally 50% water wet, Thermo Scientific Chemicals
Catalog number: 038299.06
5 g, Each
Quantity
Catalog number: 038299.06
also known as 038299-06
Price (USD)
Have Questions?
Loss on Drying
45 to 60%
Description
Palladium is used as catalysis. Palladium-on-carbon has also been used as the palladium(0) catalyst in the Suzuki reaction and Stille reaction. It is also used in jewelry, dentistry, watch making, blood sugar test strips, aircraft spark plugs and in the production of surgical instruments and electrical contacts. It is also used to make professional transverse flutes.