Thermo Scientific Chemicals
Platinum, 1% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Catalog number: 038312.14
25 g, Each
Thermo Scientific Chemicals
Platinum, 1% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Catalog number: 038312.14
25 g, Each
Quantity
Catalog number: 038312.14
also known as 038312-14
Price (USD)
Quantity
-
Have Questions?
Assay from Supplier's CofA
Metal content: 0.9-1.1%
Surface Area
900-1100 m²/g