Thermo Scientific Chemicals
Platinum Paste, Thermo Scientific Chemicals
Catalog number: 038316.14
25 g, Each
Thermo Scientific Chemicals
Platinum Paste, Thermo Scientific Chemicals
Catalog number: 038316.14
25 g, Each
Quantity
Catalog number: 038316.14
also known as 038316-14
Price (USD)
Quantity
-
Have Questions?
Assay from Supplier's CofA
2.8-3.2% metal content
Total Metal Impurities
Report (ppm)
Surface Area
900-1100 m²/g
Description
Platinum is used in catalytic converters, laboratory equipment, electrical application and electrodes, platinum resistance thermometers, dentistry equipment, and jewellery. It is also used in glass fiber applications, chemical processing, off-road vehicles and fuel cells.