Thermo Scientific Chemicals
Platinum, 10% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Catalog number: 038325.14
25 g, Each
Thermo Scientific Chemicals
Platinum, 10% on activated carbon powder, standard, reduced, nominally 50% water wet, Thermo Scientific Chemicals
Catalog number: 038325.14
25 g, Each
Quantity
Catalog number: 038325.14
also known as 038325-14
Price (USD)
Quantity
-
Have Questions?
Surface Area
900-1100 m²/g
Assay from Supplier's CofA
Metal content (Pt): 9.5-10.5%
Description
Platinum is used in catalytic converters, laboratory equipment, electrical application and electrodes, platinum resistance thermometers, dentistry equipment, and jeweler. It is also used in glass fiber applications, chemical processing, off-road vehicles and fuel cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Aromatic ring hydrogenation, aliphatic aldehydes and ketones to alcohols, conversion of -CH=CH- to -CH2-CH2-