Thermo Scientific Chemicals

Cadmium sulfide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 038342.18
50 g, Each
Thermo Scientific Chemicals

Cadmium sulfide, 99.999% (metals basis), Thermo Scientific Chemicals

Catalog number: 038342.18
50 g, Each
Quantity
Catalog number: 038342.18
also known as 038342-18
Price (USD)
Have Questions?

Chemical Identifiers

CAS
1306-23-6
IUPAC Name
cadmium(2+) sulfanediide
Molecular Formula
CdS
InChI Key
FRLJSGOEGLARCA-UHFFFAOYSA-N
SMILES
[S--].[Cd++]
Total Metal Impurities
0.001% max.

Description

The main application of cadmium sulfide is as a pigment. Cadmium sulfide is also used in the production of solar cells. Used as light dependent resistors/photo resistors for light sensors. Used in MOCVD, sputtering and electrochemical deposition. In screen printing, it is used as a slurry containing dispersed cadmium sulfide. It is also used in dandruff shampoos.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Cancer – www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text