Chemical Name or MaterialSodium chloride crystal optic disc
Name Note25mm x 5mm, Unpolished
Recommended StorageAmbient temperatures
Quantity1 Ea.
EINECS Number231-598-3
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.