Calcium fluoride crystal optic disc, 22mm x 4mm, polished both sides, Thermo Scientific Chemicals
Calcium fluoride crystal optic disc, 22mm x 4mm, polished both sides, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium fluoride crystal optic disc, 22mm x 4mm, polished both sides, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 Ea.
Catalog number 038429.KT
Price (USD)/ Each
178.00
-
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)/ Each
178.00
Add to cart
Calcium fluoride crystal optic disc, 22mm x 4mm, polished both sides, Thermo Scientific Chemicals
Catalog number038429.KT
Price (USD)/ Each
178.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialCalcium fluoride crystal optic disc
Name Note22mm x 4mm, Polished both sides
Recommended StorageAmbient temperatures
Quantity1 Ea.
EINECS Number232-188-7
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only