Calcium fluoride crystal optic disk, 25mm x 4mm, polished both sides, Thermo Scientific Chemicals
Calcium fluoride crystal optic disk, 25mm x 4mm, polished both sides, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Calcium fluoride crystal optic disk, 25mm x 4mm, polished both sides, Thermo Scientific Chemicals

 
Have Questions?
Quantity:
1 Ea.
Catalog number 038430.KT
Price (USD)/ Each
322.00
-
Add to cart
Quantity:
1 Ea.
Request bulk or custom format
Price (USD)/ Each
322.00
Add to cart
Calcium fluoride crystal optic disk, 25mm x 4mm, polished both sides, Thermo Scientific Chemicals
Catalog number038430.KT
Price (USD)/ Each
322.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialCalcium fluoride Crystal Optic Disk
Name NotePolished on both sides
Health Hazard 1H301-H332-H315-H319-H335: Toxic if swallowed. Harmful if inhaled. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation.
Recommended StorageAmbient temperatures
Quantity1 Ea.
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only