Thermo Scientific Chemicals

Lanthanum Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, 100 Orientation, Thermo Scientific Chemicals

Catalog number: 038501.KU
5 Ea., Each
Thermo Scientific Chemicals

Lanthanum Aluminum Oxide Substrate, 10 x 10 x 0.5mm, Polished One Side, 100 Orientation, Thermo Scientific Chemicals

Catalog number: 038501.KU
5 Ea., Each
Quantity
Catalog number: 038501.KU
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12003-65-5
Appearance (Color)
White to pale cream
Comment
Orientation = 100
Form
Crystals
Particle Size
10x10x0.5mm, polished one side

Description

Used in epitaxial thin films, non-epitaxial thin films and substrate. Mainly used in making optical glass parts and optical fibers for electronic industry. Also often used in ceramic industry and catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text