Thermo Scientific Chemicals
Magnesium oxide substrate, 10x10x1mm, polished one side, 100 orientation, Thermo Scientific Chemicals
Catalog number: 038503.KT
1 Ea., Each
Thermo Scientific Chemicals
Magnesium oxide substrate, 10x10x1mm, polished one side, 100 orientation, Thermo Scientific Chemicals
Catalog number: 038503.KT
1 Ea., Each
Quantity
Catalog number: 038503.KT
also known as 038503-KT
Price (USD)
Quantity
-
Have Questions?
Description
Magnesium oxide substrate is used widely in high temperature superconductor. It is also used as deposition of ferro electric thin film coatings on magnesium oxide substrates. It is also growing applications for MgO in the field of plasma display panel (PDP) technology.