Thermo Scientific Chemicals

Strontium titanium oxide substrate, 10x10x0.5mm, polished one side, 100 orientation, Thermo Scientific Chemicals

Catalog number: 038506.KT
1 Ea., Each
Thermo Scientific Chemicals

Strontium titanium oxide substrate, 10x10x0.5mm, polished one side, 100 orientation, Thermo Scientific Chemicals

Catalog number: 038506.KT
1 Ea., Each
Quantity
Catalog number: 038506.KT
also known as 038506-KT
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12060-59-2
Form
Crystals
Particle Size
10x10x0.5mm, polished one side
Appearance (Color)
White
Comment
Orientation = 100

Description

Strontium titanium oxide substrate is used as diamond stimulant, in precision optics, in varistors, and in advanced ceramics. It is also used in in radioisotope thermoelectric generators.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text