Thermo Scientific Chemicals

Tetramethylcyclopentadiene, mixed isomers, tech. 85%, Thermo Scientific Chemicals

Catalog number: 038577.06
5 g, Each
Thermo Scientific Chemicals

Tetramethylcyclopentadiene, mixed isomers, tech. 85%, Thermo Scientific Chemicals

Catalog number: 038577.06
5 g, Each
Quantity
Catalog number: 038577.06
also known as 038577-06
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
4249-10-9
IUPAC Name
1,2,3,4-tetramethylcyclopenta-1,3-diene
Molecular Formula
C9H14
InChI Key
VNPQQEYMXYCAEZ-UHFFFAOYSA-N
SMILES
CC1=C(C)C(C)=C(C)C1
Appearance (Color)
Colorless to yellow to orange
Form
Liquid
Assay (GC)
≥80.0% (mixture of isomers)
Refractive Index
1.4700-1.4760 @ 20°C

Description

Tetramethylcyclopentadiene is used in pharmaceutical intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text