Cerium(IV) fluoride hydrate, Thermo Scientific Chemicals
Cerium(IV) fluoride hydrate, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Cerium(IV) fluoride hydrate, Thermo Scientific Chemicals

Have Questions?
Quantity:
10 g
Catalog number 038580.09
also known as 038580-09
Price (USD)/ Each
91.20
-
Add to cart
Quantity:
10 g
Request bulk or custom format
Price (USD)/ Each
91.20
Add to cart
Cerium(IV) fluoride hydrate, Thermo Scientific Chemicals
Catalog number038580.09
Price (USD)/ Each
91.20
-
Add to cart
Chemical Identifiers
CAS60627-09-0
IUPAC Nametetrafluorocerium
Molecular FormulaCeF4
InChI KeySWURHZJFFJEBEE-UHFFFAOYSA-J
SMILESF[Ce](F)(F)F
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (unspecified)F 29% min
Cerium(IV) fluoride hydrates a water insoluble Cerium source for use in oxygen-sensitive applications, such as metal production. Commercial applications for cerium include metallurgy, glass and glass polishing, ceramics, catalysts, and in phosphors. In steel manufacturing it is used to remove free oxygen and sulfur by forming stable oxysulfides and by tying up undesirable trace elements, such as lead and antimony.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only