(2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium, 98%, Thermo Scientific Chemicals
(2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

(2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
5 g
Catalog number 038596.06
also known as 038596-06
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
-
Add to cart
Quantity:
5 g
Request bulk or custom format
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
Add to cart
(2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium, 98%, Thermo Scientific Chemicals
Catalog number038596.06
Price (USD)/ Each
170.65
Online exclusive
189.00 
Save 18.35 (10%)
-
Add to cart
Specifications
Chemical Name or Material(2,2,6,6-Tetramethyl-3,5-heptanedionato)lithium
CAS22441-13-0
Health Hazard 1H315-H319-H335
Health Hazard 2GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
View more
Used as reactant for synthesis of Bis-cycloiridiated complexes for electroluminescent studies, cationic imido alkylidene complexes, polyaromatic ligands containing pyrene and mono-cyclometalated platinum complexes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only