Bis(pentamethylcyclopentadienyl)manganese, 98%, Thermo Scientific Chemicals
Bis(pentamethylcyclopentadienyl)manganese, 98%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Bis(pentamethylcyclopentadienyl)manganese, 98%, Thermo Scientific Chemicals

Have Questions?
Quantity:
1 g
Catalog number 038605.03
also known as 038605-03
Price (USD)/ Each
118.00
-
Add to cart
Quantity:
1 g
Request bulk or custom format
Price (USD)/ Each
118.00
Add to cart
Bis(pentamethylcyclopentadienyl)manganese, 98%, Thermo Scientific Chemicals
Catalog number038605.03
Price (USD)/ Each
118.00
-
Add to cart
Chemical Identifiers
CAS67506-86-9
Molecular FormulaC20H30Mn
InChI KeyDBNMEMKBUCQRTO-UHFFFAOYSA-N
SMILESCc1c(C)c(C)[c-](C)c1C.C[C-]12[C-]3(C)[C-]4(C)[C-]5(C)[C-]1(C)[Mn]2345
Molecular Weight (g/mol)325.40
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Orange to red-orange to red
FormCrystals or crystalline powder
Elemental AnalysisCarbon (C): ≥71.9%
Elemental AnalysisHydrogen (H): ≥9.0%
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only