Manganese(II) 2-ethylhexanoate, 40% w/w in mineral spirits, 6% Mn, Thermo Scientific Chemicals
Manganese(II) 2-ethylhexanoate, 40% w/w in mineral spirits, 6% Mn, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Manganese(II) 2-ethylhexanoate, 40% w/w in mineral spirits, 6% Mn, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
250 g
5 kg
Catalog number 038607.30
also known as 038607-30
Price (USD)/ Each
28.60
-
Add to cart
Quantity:
250 g
Request bulk or custom format
Price (USD)/ Each
28.60
Add to cart
Manganese(II) 2-ethylhexanoate, 40% w/w in mineral spirits, 6% Mn, Thermo Scientific Chemicals
Catalog number038607.30
Price (USD)/ Each
28.60
-
Add to cart
Chemical Identifiers
CAS13434-24-7
IUPAC Namemanganese(2+) bis(2-ethylhexanoate)
Molecular FormulaC16H30MnO4
InChI KeyFHRAKXJVEOBCBQ-UHFFFAOYNA-L
SMILES[Mn++].CCCCC(CC)C([O-])=O.CCCCC(CC)C([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Specific Gravity0.870-0.917 @ 20?C
Residue on Evaporation32.00-42.00%
ViscosityA5 - A (Gardner Holdt @ 25?C)
Elemental AnalysisMn: 5.90-6.10%
Used as paint driers. Accelerating curing agent for unsaturated polyester. Also used as a intermediate for dyestuff, pharmaceutical, syntheses material.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only