Thermo Scientific Chemicals

Barium fluoride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 038610.18
50 g, Each
Thermo Scientific Chemicals

Barium fluoride, ultra dry, 99.998% (metals basis), Thermo Scientific Chemicals

Catalog number: 038610.18
50 g, Each
Quantity
Catalog number: 038610.18
also known as 038610-18
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
7787-32-8
IUPAC Name
barium(2+) difluoride
Molecular Formula
BaF2
InChI Key
OYLGJCQECKOTOL-UHFFFAOYSA-L
SMILES
[F-].[F-].[Ba++]
Appearance (Color)
White
Form
Powder
Residual water
< 100 ppm
Total Metal Impurities
0.002% max.

Description

Barium Fluoride is used in ceramic flux, carbon brushes for electrical equipment, glass making, in the manufacture of other fluorides, crystals for spectroscopy, electronics and dry-film lubricants. Barium Fluoride is used in spectroscopic components. Barium Fluoride is often suitable for applications in the passive IR band (8 to 14 ?m) and is often used as a viewport window for thermography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

  • Ampouled under argon
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text