Thermo Scientific Chemicals

Bromophenol Blue sodium salt, 0.04% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: 038693.AE
100 mL, Each
Thermo Scientific Chemicals

Bromophenol Blue sodium salt, 0.04% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: 038693.AE
100 mL, Each
Quantity
Catalog number: 038693.AE
also known as 038693-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34725-61-6
IUPAC Name
sodium 3,5-dibromo-4-[3-(2,6-dibromo-4-hydroxyphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]benzen-1-olate
Molecular Formula
C19H9Br4NaO5S
InChI Key
KGANHHDZDYZJEM-UHFFFAOYNA-M
SMILES
[Na+].OC1=CC(Br)=C(C(Br)=C1)C1(OS(=O)(=O)C2=CC=CC=C12)C1=C(Br)C=C([O-])C=C1Br
Form
Liquid
Assay from Supplier's CofA
0.040% w/v ? 0.002% w/v
Assay from Supplier's CofA
pH 3.0 (yellow) to pH 4.6 (blue)

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text