Thermo Scientific Chemicals

Methyl Orange, 0.1% w/v aq. soln.

Catalog number: 038695.AP
500 mL, Each
Thermo Scientific Chemicals

Methyl Orange, 0.1% w/v aq. soln.

Catalog number: 038695.AP
500 mL, Each
Quantity
Catalog number: 038695.AP
also known as 038695-AP
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
547-58-0
IUPAC Name
4-{2-[4-(dimethylamino)phenyl]diazen-1-yl}benzene-1-sulfonate
Molecular Formula
C14H14N3O3S
InChI Key
IETWCRRCPURZOC-UHFFFAOYSA-M
SMILES
CN(C)C1=CC=C(C=C1)N=NC1=CC=C(C=C1)S([O-])(=O)=O
Form
Liquid
Assay from Supplier's CofA
0.100% w/v ? 0.005% w/v
Assay from Supplier's CofA
pH Range: pH 3.2 (Red) to 4.4 (Yellow)
Appearance (Color)
Orange

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text