Thermo Scientific Chemicals

Bromocresol Green sodium salt, 0.04% w/v aq. soln.

Catalog number: 038696.AE
100 mL, Each
Thermo Scientific Chemicals

Bromocresol Green sodium salt, 0.04% w/v aq. soln.

Catalog number: 038696.AE
100 mL, Each
Quantity
Catalog number: 038696.AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62625-32-5
IUPAC Name
sodium 2,6-dibromo-4-[3-(3,5-dibromo-4-hydroxy-2-methylphenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-3-methylbenzen-1-olate
Molecular Formula
C21H13Br4NaO5S
InChI Key
HEFSGAHJDGZCHA-UHFFFAOYNA-M
SMILES
[Na+].CC1=C(Br)C(O)=C(Br)C=C1C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C([O-])C(Br)=C1C
Appearance (Color)
Green to green-blue to blue
Form
Liquid
Assay from Suppliers CofA
0.040% w/v ± 0.002% w/v
Comment
Green/yellow at pH 3.8, Blue at pH 5.4

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text