Thermo Scientific Chemicals

Bromocresol Purple sodium salt, 0.04% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: 038700.AE
100 mL, Each
Thermo Scientific Chemicals

Bromocresol Purple sodium salt, 0.04% w/v aq. soln., Thermo Scientific Chemicals

Catalog number: 038700.AE
100 mL, Each
Quantity
Catalog number: 038700.AE
also known as 038700-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
62625-30-3
IUPAC Name
2-[(3-bromo-4-hydroxy-5-methylphenyl)[(1E)-3-bromo-5-methyl-4-oxocyclohexa-2,5-dien-1-ylidene]methyl]benzene-1-sulfonate
Molecular Formula
C21H15Br2O5S
InChI Key
HJZFRHKAXRLQSG-XMHGGMMESA-M
SMILES
CC1=CC(=CC(Br)=C1O)C(=C1\C=C(C)C(=O)C(Br)=C1)\C1=CC=CC=C1S([O-])(=O)=O
Transition Interval
pH 5.2 (yellow-green) to pH 6.8 (purple)
Concentration (w/v)
0.04% w/v aq. soln

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text