Thermo Scientific Chemicals

Bromothymol Blue sodium salt, 0.04% w/v aqueous solution, Thermo Scientific Chemicals

Catalog number: 038701.AE
100 mL, Each
Thermo Scientific Chemicals

Bromothymol Blue sodium salt, 0.04% w/v aqueous solution, Thermo Scientific Chemicals

Catalog number: 038701.AE
100 mL, Each
Quantity
Catalog number: 038701.AE
also known as 038701-AE
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
34722-90-2
IUPAC Name
sodium 2-{[(1Z)-3-bromo-2-methyl-4-oxo-5-(propan-2-yl)cyclohexa-2,5-dien-1-ylidene][3-bromo-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl]methyl}benzene-1-sulfonate
Molecular Formula
C27H27Br2NaO5S
InChI Key
NMKFVGALBGZKGW-FKWCIMQXSA-M
SMILES
[Na+].CC(C)C1=CC(\C(C2=CC=CC=C2S([O-])(=O)=O)=C2/C=C(C(C)C)C(=O)C(Br)=C2C)=C(C)C(Br)=C1O
Transition Interval
pH 6.0 (yellow) to pH 7.6 (blue)
Concentration (w/v)
Bromothymol Blue sodium salt, 0.04% w/v aq. soln.

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text