Thermo Scientific Chemicals

Phenolphthalein, 0.5% w/v in alcohol

Catalog number: 038703.AP
500 mL, Each
Thermo Scientific Chemicals

Phenolphthalein, 0.5% w/v in alcohol

Catalog number: 038703.AP
500 mL, Each
Quantity
Catalog number: 038703.AP
also known as 038703-AP
Price (USD)
Have Questions?

Chemical Identifiers

CAS
77-09-8
IUPAC Name
3,3-bis(4-hydroxyphenyl)-1,3-dihydro-2-benzofuran-1-one
Molecular Formula
C20H14O4
InChI Key
KJFMBFZCATUALV-UHFFFAOYSA-N
SMILES
OC1=CC=C(C=C1)C1(OC(=O)C2=CC=CC=C12)C1=CC=C(O)C=C1
Appearance (Color)
Clear colorless
Concentration (w/v)
0.5% w/v in alcohol
Transition Interval
pH 8 colorless to pH 10 red
Form
Liquid

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
WARNING: Cancer and Reproductive Harm – www.P65Warnings.ca.gov
For Manufacturing and Laboratory Use. Not for beverage use

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text