Thermo Scientific Chemicals

Tropaeolin O

Catalog number: 038708.09
10 g, Each
Thermo Scientific Chemicals

Tropaeolin O

Catalog number: 038708.09
10 g, Each
Quantity
Catalog number: 038708.09
also known as 038708-09
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
547-57-9
IUPAC Name
4-[2-(2-hydroxy-4-oxocyclohexa-2,5-dien-1-ylidene)hydrazin-1-yl]benzene-1-sulfonic acid sodium
Molecular Formula
C12H10N2NaO5S
InChI Key
YOGUPPQYRWYKGT-UHFFFAOYSA-N
SMILES
[Na].OC1=CC(=O)C=CC1=NNC1=CC=C(C=C1)S(O)(=O)=O
Appearance (Color)
Orange to brown

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text