Buffer solution pH 10.00 (+/-0.024 @ 25oC) Colored Blue Specpure NIST Traceable, Thermo Scientific Chemicals
Buffer solution pH 10.00 (+/-0.024 @ 25oC) Colored Blue Specpure NIST Traceable, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Buffer solution pH 10.00 (+/-0.024 @ 25oC) Colored Blue Specpure NIST Traceable, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
1 L
Catalog number 038709.K7
also known as 038709-K7
Price (USD)/ Each
73.65
Online exclusive
82.10 
Save 8.45 (10%)
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
73.65
Online exclusive
82.10 
Save 8.45 (10%)
Add to cart
Buffer solution pH 10.00 (+/-0.024 @ 25oC) Colored Blue Specpure NIST Traceable, Thermo Scientific Chemicals
Catalog number038709.K7
Price (USD)/ Each
73.65
Online exclusive
82.10 
Save 8.45 (10%)
-
Add to cart
Chemical Identifiers
CAS1310-73-2, 7778-77-0, 7732-18-5
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofApH 10.00 (± 0.024 @ 25°C)
Appearance (Color)Blue
Buffer solution (pH 10.00) is used as a means of keeping pH at a nearly constant value in different chemical applications. Buffer solution is used in alcoholic fermentation, leather tanning, electroplating, organic synthesis, sugar and paper manufacturing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only