Buffer solution pH 10.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
Buffer solution pH 10.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Buffer solution pH 10.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 038710.AP
also known as 038710-AP
Price (USD)/ Each
27.65
Online exclusive
30.90 
Save 3.25 (11%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
27.65
Online exclusive
30.90 
Save 3.25 (11%)
Add to cart
Buffer solution pH 10.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
Catalog number038710.AP
Price (USD)/ Each
27.65
Online exclusive
30.90 
Save 3.25 (11%)
-
Add to cart
Chemical Identifiers
CAS1336-21-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay from Supplier's CofApH 10.00 (± 0.024 @ 25°C)
FormLiquid
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only