Thermo Scientific Chemicals

Buffer solution pH 7.00 (+/-0.024 @ 25oC) Colored Green Specpure NIST Traceable, Thermo Scientific Chemicals

Catalog number: 038711.K7
4 L, Each
Thermo Scientific Chemicals

Buffer solution pH 7.00 (+/-0.024 @ 25oC) Colored Green Specpure NIST Traceable, Thermo Scientific Chemicals

Catalog number: 038711.K7
4 L, Each
Quantity
Catalog number: 038711.K7
also known as 038711-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1336-21-6
Appearance (Color)
Green
Assay from Supplier's CofA
pH 7.00 (± 0.024 at 25°C)
Form
Liquid

Description

Buffer solutions play a substantial role in industrial and physiological processes. Buffer solutions of known and precise pH value allow to set the testing system to measure the samples of unknown pH more precisely and accurately.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text