Buffer solution pH 7.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
Buffer solution pH 7.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Buffer solution pH 7.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
1 L
4 L
Catalog number 038712.AP
also known as 038712-AP
Price (USD)/ Each
28.65
Online exclusive
31.30 
Save 2.65 (8%)
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
28.65
Online exclusive
31.30 
Save 2.65 (8%)
Add to cart
Buffer solution pH 7.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
Catalog number038712.AP
Price (USD)/ Each
28.65
Online exclusive
31.30 
Save 2.65 (8%)
-
Add to cart
Chemical Identifiers
CAS1336-21-6
SpecificationsSpecification SheetSpecification Sheet
Assay from Supplier's CofApH 7.00
Appearance (Color)Colorless
FormLiquid
Used to determine pH of solutions in various industrial applications. Buffer solutions of known and accurate pH value allows to adjust testing system to measure the samples with unknown pH more precisely and accurately.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only