Thermo Scientific Chemicals

Buffer solution pH 7.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals

Catalog number: 038712.K2
1 L, Each
Thermo Scientific Chemicals

Buffer solution pH 7.00 (+/-0.024 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals

Catalog number: 038712.K2
1 L, Each
Quantity
Catalog number: 038712.K2
also known as 038712-K2
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
1336-21-6
Assay from Supplier's CofA
pH 7.00
Appearance (Color)
Colorless
Form
Liquid

Description

Used to determine pH of solutions in various industrial applications. Buffer solutions of known and accurate pH value allows to adjust testing system to measure the samples with unknown pH more precisely and accurately.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text