Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
500 mL
1 L
Catalog number 038714.K7
also known as 038714-K7
Price (USD)/ Each
81.60
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
81.60
Add to cart
Buffer solution pH 4.01 (+/-0.022 @ 25oC) No Color Specpure NIST Traceable, Thermo Scientific Chemicals
Catalog number038714.K7
Price (USD)/ Each
81.60
-
Add to cart
Chemical Identifiers
CAS1336-21-6
SpecificationsSpecification SheetSpecification Sheet
FormLiquid
Assay from Supplier's CofApH 4.01 (± 0.022 @ 25°C)
Appearance (Color)Clear colorless
Buffer solutions are used as a means of keeping pH at a nearly constant value in a wide variety of chemical applications. Buffer solutions are essential to maintain the suitable condition to keep the correct pH for enzymes in many organisms to work. It is also used in fermentation processes and in setting the correct conditions for dyes used in coloring fabrics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only