Aluminum, plasma standard solution, Specpure™, Al 1000 mg/ml, Thermo Scientific Chemicals
Aluminum, plasma standard solution, Specpure™, Al 1000 mg/ml, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Aluminum, plasma standard solution, Specpure™, Al 1000 mg/ml, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
50 mL
100 mL
Catalog number 038727.AP
also known as 038727-AP
Price (USD)/ Each
167.00
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)/ Each
167.00
Add to cart
Aluminum, plasma standard solution, Specpure™, Al 1000 mg/ml, Thermo Scientific Chemicals
Catalog number038727.AP
Price (USD)/ Each
167.00
-
Add to cart
SpecificationsSpecification SheetSpecification Sheet
Chemical Name or MaterialAluminum
Name Noteplasma standard solution, Al 1000μg/mL
CAS7784-27-2
Health Hazard 1H290-H314-H335
Quantity500 mL
View more
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only