Thermo Scientific Chemicals
Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 038728.AD
50 mL, Each
Thermo Scientific Chemicals
Chromium, plasma standard solution, Specpure™ Cr 1000μg/mL, Thermo Scientific Chemicals
Catalog number: 038728.AD
50 mL, Each
Quantity
Catalog number: 038728.AD
also known as 038728-AD
Price (USD)
Quantity
-
Have Questions?
Concentration
1,000?g/mL ?1%
Starting Material
Cr(NO3)3