Hydrofluoric acid, Environmental Grade Plus, 47-51%, Thermo Scientific Chemicals
Hydrofluoric acid, Environmental Grade Plus, 47-51%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Hydrofluoric acid, Environmental Grade Plus, 47-51%, Thermo Scientific Chemicals

Have Questions?
Quantity:
500 mL
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog number 038746.AP
also known as 038746-AP
Price (USD)/ Each
-
Quantity:
500 mL
Request bulk or custom format
Chemical Identifiers
CAS7664-39-3
IUPAC Namehydrogen fluoride
Molecular FormulaFH
InChI KeyKRHYYFGTRYWZRS-UHFFFAOYSA-N
SMILESF
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
Assay from Supplier's CofA47.0-51.0%
FormLiquid
Total Metal Impurities≤1.0 ppm
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only