Sulfuric acid has a strong dehydrating agent and is applied in many applications like the domestic acidic drain cleaner, electrolyte in lead-acid batteries and in various other cleansing agents. It is also useful for mineral processing, fertilizer manufacturing, petroleum purification, wastewater processing, and chemical synthesis. The catalytic decomposition of sulfuric acid when mixed with Cr/Fe oxides, shows a useful application in the sulfur iodine cycle for hydrogen production.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.