Thermo Scientific Chemicals

Petroleum ether 35/60, Thermo Scientific Chemicals

Catalog number: 038985.K7
4 L, Each
Thermo Scientific Chemicals

Petroleum ether 35/60, Thermo Scientific Chemicals

Catalog number: 038985.K7
4 L, Each
Quantity
Catalog number: 038985.K7
also known as 038985-K7
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
8032-32-4
Residue on Evaporation
≤5ppm
Comment
Suitable for liquid chromatography, residue analysis.
Impurity content
Chlorinated pesticides ≤5ppt
Comment
Filtered through 0.2?M filters.
Residual water
≤0.03%

Description

Petroleum ether 35/60 is used as a mobile phase in High Performance Liquid Chromatography and Liquid Chromatography coupled with Mass Spectrometry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text