Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, HPLC Grade, 99.5%, Thermo Scientific Chemicals

Catalog number: 038986.K7
4 L, Each
Thermo Scientific Chemicals

1-Methyl-2-pyrrolidinone, HPLC Grade, 99.5%, Thermo Scientific Chemicals

Catalog number: 038986.K7
4 L, Each
Quantity
Catalog number: 038986.K7
also known as 038986-K7
Price (USD)
Have Questions?

Chemical Identifiers

CAS
872-50-4
IUPAC Name
1-methylpyrrolidin-2-one
Molecular Formula
C5H9NO
InChI Key
SECXISVLQFMRJM-UHFFFAOYSA-N
SMILES
CN1CCCC1=O
Optical Absorbance at
285 nm: ≤1.00, 300 nm: ≤ 0.5 nm, 325 nm: ≤0.10 nm, 350 nm: ≤0.03, 400 nm: ≤0.01 AU
Residual water
≤0.05%
Form
Liquid
Assay from Suppliers CofA
≥99.5%
Residue on ignition
5ppm

Description

1-methyl-2-pyrrolidinone, is useful for spectrophotometry, chromatography and ICP-MS detection.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text