Glycerol, ultrapure, HPLC Grade, Thermo Scientific Chemicals
Glycerol, ultrapure, HPLC Grade, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Glycerol, ultrapure, HPLC Grade, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
4 L
500 mL
1 L
4 x 1 L
Catalog number 038988.K7
also known as 038988-K7
Price (USD)/ Each
347.00
-
Add to cart
Quantity:
4 L
Request bulk or custom format
Price (USD)/ Each
347.00
Add to cart
Glycerol, ultrapure, HPLC Grade, Thermo Scientific Chemicals
Catalog number038988.K7
Price (USD)/ Each
347.00
-
Add to cart
Chemical Identifiers
CAS56-81-5
IUPAC Namepropane-1,2,3-triol
Molecular FormulaC3H8O3
InChI KeyPEDCQBHIVMGVHV-UHFFFAOYSA-N
SMILESOCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Assay (GC)99.0%, min
Optical Absorbance at206 nm 1.00 AU, max
Optical Absorbance at225 nm 0.25 AU, max
Optical Absorbance at320 nm 0.02 AU, max
Optical Absorbance at210 nm 0.70 AU, max
View more
Glycerol is used as emulsifier, thickening agent in cosmetics and foods, plasticizer in packaging, paints and paper industry, preservative in food and beverages. It is used in pharmaceutical preparations to improve smoothness and providing lubrication. It is also used in epichlorohydrin and mono propylene glycol production.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only