Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Name or Material | Diethyl ether |
Melting Point | °C to 116°C |
CAS | 60-29-7 |
Health Hazard 1 | H224-H302-H305-H335-H336-H373 |
Health Hazard 3 | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P314-P330-P370+P378q-P501c |
Recommended Storage | Ambient temperatures |
DOT Information | Hazard Class: 3; Packaging Group: I |
Density | 0.713 g/mL |
RTECS Number | KI5775000 |
Boiling Point | 34.6°C |
EINECS Number | 200-467-2 |
Quantity | 4 x 1 L |
TSCA | Yes |
Flash Point | -45°C (-49°F) |
Physical Form | Liquid |
Assay Percent Range | 99% |
Molecular Formula | C4H10O |
MDL Number | MFCD00011646 |
Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
Solubility Information | Immiscible with water,miscible with alcohol. |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
SMILES | CCOCC |
IUPAC Name | ethoxyethane |
Molecular Weight (g/mol) | 74.12 |
PubChem CID | 3283 |
ChEBI | CHEBI:35702 |
Formula Weight | 74.12 |
Grade | HPLC |
Unit Size | Each |
Chemical Name or Material | Diethyl ether |
Melting Point | °C to 116°C |
CAS | 60-29-7 |
Health Hazard 1 | H224-H302-H305-H335-H336-H373 |
Health Hazard 3 | P210-P233-P235-P240-P241-P242-P243-P260-P264b-P270-P271-P280-P301+P312-P303+P361+P353-P304+P340-P312-P314-P330-P370+P378q-P501c |
Recommended Storage | Ambient temperatures |
DOT Information | Hazard Class: 3; Packaging Group: I |
Density | 0.713 g/mL |
RTECS Number | KI5775000 |
Boiling Point | 34.6°C |
EINECS Number | 200-467-2 |
Quantity | 1 L |
TSCA | Yes |
Flash Point | -45°C (-49°F) |
Physical Form | Liquid |
Assay Percent Range | 99% |
Molecular Formula | C4H10O |
MDL Number | MFCD00011646 |
Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
Solubility Information | Immiscible with water,miscible with alcohol. |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
SMILES | CCOCC |
IUPAC Name | ethoxyethane |
Molecular Weight (g/mol) | 74.12 |
PubChem CID | 3283 |
ChEBI | CHEBI:35702 |
Formula Weight | 74.12 |
Grade | HPLC |
Unit Size | Each |
Chemical Name or Material | Diethyl ether |
CAS | 60-29-7 |
Density | 0.713g/mL |
EINECS Number | 200-467-2 |
Quantity | 4 L |
Physical Form | Liquid |
Assay Percent Range | 99% |
Molecular Formula | C4H10O |
MDL Number | MFCD00011646 |
Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
SMILES | CCOCC |
IUPAC Name | ethoxyethane |
Molecular Weight (g/mol) | 74.12 |
PubChem CID | 3283 |
ChEBI | CHEBI:35702 |
Formula Weight | 74.12 |
Unit Size | Each |
Chemical Name or Material | Diethyl ether |
Melting Point | -116°C |
CAS | 60-29-7 |
Boiling Point | 34.6°C |
Quantity | 4 x 4 L |
TSCA | Yes |
Flash Point | −45°C |
Packaging | Glass Bottle |
Physical Form | Liquid |
Specific Gravity | 0.713 |
Molecular Formula | C4H10O |
MDL Number | MFCD00011646 |
UN Number | UN1155 |
Synonym | diethyl ether, ether, ethyl ether, diethyl oxide, ethyl oxide, aether, pronarcol, anesthetic ether, 3-oxapentane, anaesthetic ether |
InChI Key | RTZKZFJDLAIYFH-UHFFFAOYSA-N |
SMILES | CCOCC |
IUPAC Name | ethoxyethane |
Molecular Weight (g/mol) | 74.12 |
PubChem CID | 3283 |
ChEBI | CHEBI:35702 |
Formula Weight | 74.12 |
Percent Purity | 99% |
Grade | HPLC |
Unit Size | Each |
Catalog Number | Specifications | Unit Size | Quantity | Price (USD) | Availability | Quantity | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|---|
038990.M1 also known as 038990-M1 | Each | 4 x 1 L | 234.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
038990.K2 also known as 038990-K2 | Each | 1 L | 82.40 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
038990.K7 also known as 038990-K7 | Each | 4 L | 163.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
| |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
038990.M6 also known as 038990-M6 | Each | 4 x 4 L | 544.00 | - | |||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||||
|
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.