Carbon disulfide, HPLC Grade, 99.8+%, Thermo Scientific Chemicals
Carbon disulfide, HPLC Grade, 99.8+%, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Carbon disulfide, HPLC Grade, 99.8+%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
4 x 1 L
Catalog number 038993.K2
also known as 038993-K2
Price (USD)/ Each
112.65
Online exclusive
125.00 
Save 12.35 (10%)
-
Add to cart
Quantity:
1 L
Request bulk or custom format
Price (USD)/ Each
112.65
Online exclusive
125.00 
Save 12.35 (10%)
Add to cart
Carbon disulfide, HPLC Grade, 99.8+%, Thermo Scientific Chemicals
Catalog number038993.K2
Price (USD)/ Each
112.65
Online exclusive
125.00 
Save 12.35 (10%)
-
Add to cart
Chemical Identifiers
CAS75-15-0
IUPAC Namemethanedithione
Molecular FormulaCS2
InChI KeyQGJOPFRUJISHPQ-UHFFFAOYSA-N
SMILESS=C=S
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
FormLiquid
Color (APHA)≤10
Assay (GC)≥99.8%
CommentSpecification differs for U.S. and non-U.S. material where indicated
View more
Carbon disulfide is used in spectrophotometry, high performance liquid chromatography and environmental testing.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

WARNING: Reproductive Harm - www.P65Warnings.ca.gov
RUO – Research Use Only