Thermo Scientific Chemicals

Titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 039000.14
25 g, Each
Thermo Scientific Chemicals

Titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals

Catalog number: 039000.14
25 g, Each
Quantity
Catalog number: 039000.14
also known as 039000-14
Price (USD)
Quantity
-
Have Questions?

Chemical Identifiers

CAS
12065-65-5
Total Metal Impurities
0.1% max.

Description

Nanocrystalline titanium oxide electrodes is useful for photo voltaic applications. It plays an important role as photocatalyst under visible light radiation. Treated titanium oxide nanotubes play a significant part in the development of nano-scale hydroxy apatite. It is too utilized as catalysts.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text