Titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Thermo Scientific Chemicals
Titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Useful for photo voltaic applications. It plays an important role as photocatalyst under visible light radiation. Treated titanium oxide nanotubes play a significant part in the development of nano-scale hydroxy apatite. It is too utilized as catalysts.
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Catalog number 039000.22
also known as 039000-22
Price (USD)/ Each
357.00
-
Add to cart
Quantity:
100 g
Request bulk or custom format
Price (USD)/ Each
357.00
Add to cart
Titanium oxide, 99.9% (metals basis), Thermo Scientific Chemicals
Catalog number039000.22
Price (USD)/ Each
357.00
-
Add to cart
Chemical Identifiers
CAS12065-65-5
SpecificationsSpecification SheetSpecification Sheet
Total Metal Impurities0.1% max.
Nanocrystalline titanium oxide electrodes is useful for photo voltaic applications. It plays an important role as photocatalyst under visible light radiation. Treated titanium oxide nanotubes play a significant part in the development of nano-scale hydroxy apatite. It is too utilized as catalysts.