Thermo Scientific Chemicals

Glassy Carbon Conical Crucible

Catalog number: 039003.KT
10 mL, 31mm, 27mm, Each
Thermo Scientific Chemicals

Glassy Carbon Conical Crucible

Catalog number: 039003.KT
10 mL, 31mm, 27mm, Each
Capacity (Metric)
Diameter (Metric) Outer
Height (Metric)
Catalog number: 039003.KT
also known as 039003-KT
Price (USD)
Quantity
-
Have Questions?
Form
Crucible, Glassy Carbon Conical, 10 mL volume

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Subjected to a high temperature heat treatment which imparts special material properties including significantly improved corrosion resistance and strength.

Glassy carbon offers:
  • Resistance to all wet decomposition agents
  • No memory effects (uncontrolled adsorption and release of foreign elements)
  • No contamination of analytical samples
  • Stability to acid and alkaline melts
  • No wetting effect of metal melts
  • Good resistance to thermal shock allows rapid heating and cooling times
  • Resistive to abrasive wear
  • Good electrical conductivity
RUO – Research Use Only

Figures

Documents & Downloads

Certificates

    Frequently asked questions (FAQs)

    Citations & References

    Search citations by name, author, journal title or abstract text