Lanthanum chloride, Releasing Agent Solution, Specpure™
Lanthanum chloride, Releasing Agent Solution, Specpure™
Lanthanum chloride, Releasing Agent Solution, Specpure™
Lanthanum chloride, Releasing Agent Solution, Specpure™
Thermo Scientific Chemicals

Lanthanum chloride, Releasing Agent Solution, Specpure™

CAS: 10099-58-8 | Cl3La | 245.26 g/mol
Have Questions?
Catalog NumberQuantity
039036.AP
also known as 039036-AP
500 mL
Catalog number 039036.AP
also known as 039036-AP
Price (USD)
218.65
Special offer
Online exclusive
Ends: 26-Sep-2025
270.00
Save 51.35 (19%)
Each
-
Add to cart
Quantity:
500 mL
Request bulk or custom format
Price (USD)
218.65
Special offer
Online exclusive
Ends: 26-Sep-2025
270.00
Save 51.35 (19%)
Each
Add to cart
Specifications
CAS10099-58-8
Chemical Name or MaterialLanthanum chloride
DOT InformationHazard Class: 8; Packaging Group: III
EINECS Number231-595-7
Health Hazard 1H290-H315-H319
View more
Ask our AI about this Product
Lanthanum chloride is a precursor for synthesis of lanthanum phosphate nano rods and used in gamma detectors. It is also used as a catalyst for high pressure oxidative chlorination of methane to chloromethane with hydrochloric acid and oxygen. In organic synthesis, lanthanum trichloride acts as lewis acid for the conversion of aldehydes to acetals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only
Have questions about this product? Ask our AI assisted search.
This is an AI-powered search and may not always get things right. You can help us make it better with a thumbs up or down on individual answers or by selecting the “Give feedback" button. Your search history and customer login information may be retained by Thermo Fisher and processed in accordance with our Privacy Notice.

Documents & Downloads

Certificates

Safety Data Sheets

Share catalog number, name or link