Iodine monochloride, Thermo Scientific Chemicals
Iodine monochloride, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iodine monochloride, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
100 g
Catalog number 039104.36
also known as 039104-36
Price (USD)/ Each
285.65
Online exclusive
317.00 
Save 31.35 (10%)
-
Add to cart
Quantity:
500 g
Request bulk or custom format
Price (USD)/ Each
285.65
Online exclusive
317.00 
Save 31.35 (10%)
Add to cart
Iodine monochloride, Thermo Scientific Chemicals
Catalog number039104.36
Price (USD)/ Each
285.65
Online exclusive
317.00 
Save 31.35 (10%)
-
Add to cart
Chemical Identifiers
CAS7790-99-0
IUPAC Nameiodochlorane
Molecular FormulaClI
InChI KeyQZRGKCOWNLSUDK-UHFFFAOYSA-N
SMILESClI
View more
SpecificationsSpecification SheetSpecification Sheet
Comment1.0 - 1.2 I/Cl ratio
Appearance (Color)Brown to black
FormCrystalline/Liquid
ICl is used as a catalyst in organic synthesis. It is the source of electrophilic iodine in the synthesis of certain aromatic iodides. It is used to determine the iodine value of a substance.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only