Iodine monobromide, 98% min, Thermo Scientific Chemicals
Iodine monobromide, 98% min, Thermo Scientific Chemicals
Thermo Scientific Chemicals

Iodine monobromide, 98% min, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
Catalog number 039106.14
also known as 039106-14
Price (USD)/ Each
42.65
Online exclusive
46.90 
Save 4.25 (9%)
-
Add to cart
Quantity:
25 g
Request bulk or custom format
Price (USD)/ Each
42.65
Online exclusive
46.90 
Save 4.25 (9%)
Add to cart
Iodine monobromide, 98% min, Thermo Scientific Chemicals
Catalog number039106.14
Price (USD)/ Each
42.65
Online exclusive
46.90 
Save 4.25 (9%)
-
Add to cart
Chemical Identifiers
CAS7789-33-5
IUPAC Nameiodobromane
Molecular FormulaBrI
InChI KeyCBEQRNSPHCCXSH-UHFFFAOYSA-N
SMILESBrI
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Black, grey, or blue-purple
FormCrystals or crystalline powder
Assay (unspecified)≥98.0%
Iodine bromide is used for bromination of and for determining iodine used. IBr is used in some types of iodometry. It is also used as a organic synthesis reagent.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

RUO – Research Use Only